StemPro™ hESC SFM - FAQs
StemPro™ hESC SFM - FAQs
View additional product information for StemPro™ hESC SFM - FAQs (A1000701)
11 product FAQs found
How do I prepare the StemPRO hESC SFM complete medium?
We recommend to thaw the supplement in the 37°C water bath, aliquot and freeze at -5°C to -20°C. Thaw each aliquot only one additional time. To make 100 ml complete medium, mix 90.8 ml DMEM/F-12 with GlutaMax matrix, 2 ml StemPRO hESC supplement, 7.2 ml 25% BSA solution, 80 ul bFGF (at 10 ug/ml), and 182 ul 2-mercaptoethanol (at 55mM). Store at 2°C-8°C until ready to use, but use the same day.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can StemPRO hESC SFM be used to culture mouse ESCs?
No, we don't recommend StemPro hESC SFM for mouse cells because they showed poor growth and some differentiation. We recommend KnockOut DMEM and KnockOut SR for mouse ESC cultures (catalog numbers 10829-018 and 10828-028 respectively).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can StemPRO hESC SFM be used for new cell line derivations?
Yes, derivations using StemPro hESC SFM have been carried out using either a basement membrane extract (like Geltrex growth factor) or feeder cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What cell lines has StemPRO hESC SFM been tested with?
StemPro hESC SFM has been tested with the following hESC: H1, H9, BG01, BG02, BG03, HUES6, HUES9, Cyth203, CyT49, BG01v, HES-2, HES-3, KhES-1, and TE06. The medium has also been used with the Rhesus monkey line R336.4. Some cells may require a short adaptation time but most do not.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does StemPRO hESC SFM require the use of a basement membrane matrix?
Yes, we recommend using Geltrex hESC-qualified Reduced Growth Factor basement membrane (catalog number A10480-02), CELLstart (catalog number A10142-01) or similar matrix.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What additional products do I need to make a complete StemPRO hESC SFM?
In addition to the StemPRO hESC SFM kit, you will also need the following to make a complete medium: full length recombinant human FGF (catalog # PHG0261) and 2-Mercaptoethanol (catalog # 21985-023).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the components of the StemPro hESC SFM kit?
The StemPro hESC SFM kit (Cat. No. A1000701) contains: - 500 mL DMEM/F-12 with GlutaMax (Cat. No. 10565-018)
- 10 mL StemPro hESC SFM Supplement 50X (Cat. No. A10006-01)
- 40 mL Bovine Serum Albumin 25% (BSA Solution) (Cat. No. A10008-01)
Note that the StemPRO hESC Supplement and BSA solution cannot be ordered individually.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I characterize human embryonic stem ( ES) cells?
Human ES cells are generally characterized by their typical morphology (they grow as tightly packed clusters of small cells with high ratio of nucleus to cytoplasm); surface marker expression; RT-PCR detection of stem cell-specific gene expression (such as Oct3/4, Sox2, and Nanog); alkaline phosphatase staining, and telomerase activity assay. The most commonly used ES specific surface markers include stage-specific embryonic antigens SSEA-3 and SSEA-4 for human ES cells. Other ES-specific surface antigens also include TRA-1-60 and TRA-1-81. (Science 282:1145 (1998).
How are human embryonic stem (ES) cells derived?
Human ES cells are derived from human blastocyst inner cell masses, isolated by immunosurgery with rabbit antiserum to BeWO cells (a human trophoblast cell line) (Science 282:1145 (1998)).
What are ES cells?
Embryonic stem (ES) cells are derived from the early mammalian embryo and are capable of unlimited, undifferentiated proliferation in vitro while maintaining their potential to differentiate into a wide of range of adult tissues including germ cells. The pluripotency of the ES cells is normally demonstrated in vitro by inducing ES cells to differentiate into embryoid bodies and checking lineage-specific markers for differentiated cells in three body layers (endo, meso, and ectoderm), or injecting them into immunodeficient mice and determining the cell types produced in the teratomas.
Has Gibco Geltrex Matrix been tested with StemPRO hESC serum-free Medium?
Yes, these products were designed to work together.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.