BSA BOVINE SERUM ALBUMIN 25% - FAQs
BSA BOVINE SERUM ALBUMIN 25% - FAQs
View additional product information for BSA BOVINE SERUM ALBUMIN 25% - FAQs (A1000801)
7 product FAQs found
How do I prepare the StemPRO hESC SFM complete medium?
We recommend to thaw the supplement in the 37°C water bath, aliquot and freeze at -5°C to -20°C. Thaw each aliquot only one additional time. To make 100 ml complete medium, mix 90.8 ml DMEM/F-12 with GlutaMax matrix, 2 ml StemPRO hESC supplement, 7.2 ml 25% BSA solution, 80 ul bFGF (at 10 ug/ml), and 182 ul 2-mercaptoethanol (at 55mM). Store at 2°C-8°C until ready to use, but use the same day.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can StemPRO hESC SFM be used to culture mouse ESCs?
No, we don't recommend StemPro hESC SFM for mouse cells because they showed poor growth and some differentiation. We recommend KnockOut DMEM and KnockOut SR for mouse ESC cultures (catalog numbers 10829-018 and 10828-028 respectively).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can StemPRO hESC SFM be used for new cell line derivations?
Yes, derivations using StemPro hESC SFM have been carried out using either a basement membrane extract (like Geltrex growth factor) or feeder cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What cell lines has StemPRO hESC SFM been tested with?
StemPro hESC SFM has been tested with the following hESC: H1, H9, BG01, BG02, BG03, HUES6, HUES9, Cyth203, CyT49, BG01v, HES-2, HES-3, KhES-1, and TE06. The medium has also been used with the Rhesus monkey line R336.4. Some cells may require a short adaptation time but most do not.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does StemPRO hESC SFM require the use of a basement membrane matrix?
Yes, we recommend using Geltrex hESC-qualified Reduced Growth Factor basement membrane (catalog number A10480-02), CELLstart (catalog number A10142-01) or similar matrix.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What additional products do I need to make a complete StemPRO hESC SFM?
In addition to the StemPRO hESC SFM kit, you will also need the following to make a complete medium: full length recombinant human FGF (catalog # PHG0261) and 2-Mercaptoethanol (catalog # 21985-023).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the components of the StemPro hESC SFM kit?
The StemPro hESC SFM kit (Cat. No. A1000701) contains: - 500 mL DMEM/F-12 with GlutaMax (Cat. No. 10565-018)
- 10 mL StemPro hESC SFM Supplement 50X (Cat. No. A10006-01)
- 40 mL Bovine Serum Albumin 25% (BSA Solution) (Cat. No. A10008-01)
Note that the StemPRO hESC Supplement and BSA solution cannot be ordered individually.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.