Can fluorescent protein-expressing cells be fixed?
Yes, fluorescent protein-expressing cells can be fixed using 4% paraformaldehyde in PBS for 10 min followed by one quick PBS rinse and 3 x 5 min washes with 1 mL PBS.
Search
View additional product information for TC-FlAsH™ Expression Analysis Detection Kit - Red (Fluorescent In-Gel Detection of TC-Tagged and Total Protein) - FAQs (A10068)
2 product FAQs found