2-Bromo-5-chlorobenzoic acid, 98+%
2-Bromo-5-chlorobenzoic acid, 98+%
2-Bromo-5-chlorobenzoic acid, 98+%
Thermo Scientific Chemicals

2-Bromo-5-chlorobenzoic acid, 98+%

CAS: 21739-93-5 | C7H4BrClO2 | 235.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10103.14
also known as A10103-14
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
160.00
Save 23.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
160.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS21739-93-5
IUPAC Name2-bromo-5-chlorobenzoic acid
Molecular FormulaC7H4BrClO2
InChI KeyRBCPJQQJBAQSOU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Cl)=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (HPLC)>98.0%
Assay (Aqueous acid-base Titration)>98.0 to <102.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only