2,4-Dichlorobenzonitrile, 98%
2,4-Dichlorobenzonitrile, 98%
2,4-Dichlorobenzonitrile, 98%
Thermo Scientific Chemicals

2,4-Dichlorobenzonitrile, 98%

CAS: 6574-98-7 | C7H3Cl2N | 172.008 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A10113.06
also known as A10113-06
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
62.40
Save 8.75 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
62.40
Save 8.75 (14%)
Each
Chemical Identifiers
CAS6574-98-7
IUPAC Name2,4-dichlorobenzonitrile
Molecular FormulaC7H3Cl2N
InChI KeyGRUHREVRSOOQJG-UHFFFAOYSA-N
SMILESClC1=CC=C(C#N)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)56.5-62.5?C
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.

Solubility
Soluble in Methanol. Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. V.K. Rastogi; Vaibhav Jain; M. Alcolea Palafox; D.N. Singh; R.A. Yadav. Laser Raman and IR spectra and force fields for 2,4-dichlorobenzonitrile. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2001, 57 (2), 209-216.
  2. Ibrahim Haddad; Shaun Hurley; C. S. Marvel. Poly(arylene sulfides) with pendant cyano groups as high-temperature laminating resins. Journal of Polymer Science Part A: Polymer Chemistry. 1973, 11 (11), 2793-2811.