alpha-Toluenesulfonyl chloride, 99%
alpha-Toluenesulfonyl chloride, 99%
alpha-Toluenesulfonyl chloride, 99%
Thermo Scientific Chemicals

alpha-Toluenesulfonyl chloride, 99%

CAS: 1939-99-7 | C7H7ClO2S | 190.641 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10124.06
also known as A10124-06
Price (USD)
38.65
Online Exclusive
42.60
Save 3.95 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
42.60
Save 3.95 (9%)
Each
Chemical Identifiers
CAS1939-99-7
IUPAC Namephenylmethanesulfonyl chloride
Molecular FormulaC7H7ClO2S
InChI KeyOAHKWDDSKCRNFE-UHFFFAOYSA-N
SMILESClS(=O)(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder or crystals
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Melting Point (clear melt)89-94°C
Identification (FTIR)Conforms
Appearance (Color)White to pale cream or pale pink to pink
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only