1-(2-Aminoethyl)piperazine, 98%
1-(2-Aminoethyl)piperazine, 98%
1-(2-Aminoethyl)piperazine, 98%
1-(2-Aminoethyl)piperazine, 98%
Thermo Scientific Chemicals

1-(2-Aminoethyl)piperazine, 98%

CAS: 140-31-8 | C6H15N3 | 129.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A10154.0B
also known as A10154-0B
Price (USD)
112.00
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
112.00
Each
Add to cart
Chemical Identifiers
CAS140-31-8
IUPAC Name2-(piperazin-1-yl)ethan-1-amine
Molecular FormulaC6H15N3
InChI KeyIMUDHTPIFIBORV-UHFFFAOYSA-N
SMILESNCCN1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4980-1.5020 @ 20?C
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.

Solubility
Soluble in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
RUO – Research Use Only

General References:

  1. S M Sami; R T Dorr; D S Alberts; W A Remers. 2-substituted 1,2-dihydro-3H-dibenz[de,h]isoquinoline-1,3-diones. A new class of antitumor agent. Journal of Medicinal Chemistry. 1993, 36 (6), 765-770.
  2. V T Mazaev; L B Troenkina. [Amino compound stability and transformation in an aqueous environment]. Hygiene and Sanitation. 1981, (10), 91-92.