Cadmium sulfate hydrate, 98%
Cadmium sulfate hydrate, 98%
Cadmium sulfate hydrate, 98%
Thermo Scientific Chemicals

Cadmium sulfate hydrate, 98%

CAS: 15244-35-6 | CdO4S | 208.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10168.36
also known as A10168-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS15244-35-6
IUPAC Namecadmium(2+) sulfate
Molecular FormulaCdO4S
InChI KeyQCUOBSQYDGUHHT-UHFFFAOYSA-L
SMILES[Cd++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder or powder
Complexometric Titration≥97.5 to ≤102.5% (Dry wt. basis)
Water Content (Karl Fischer Titration)≥15.0% to ≤40.8% (ca. 2-8 waters)
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

Solubility
Soluble in water (1130g/L ).

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. M. N. Papadopoulos; W. F. Giauque. Cadmium Sulfate and its Hydrates. Heat Capacities and Heats of Hydration. Application of the Third Law of Thermodynamics1. J. Am. Chem. Soc. 1955, 77,(10), 2740-2744.
  2. P. DuBois Murphy; B. C. Gerstein. Cadmium Sulfate and its Hydrates. Principal components of the cadmium-113 shielding tensors in cadmium sulfate hydrates: a nuclear magnetic resonance study of cadmium coordinated with oxygen. J. Am. Chem. Soc. 1981, 103,(12), 3282-3286.