Alexa Fluor™ 350 NHS Ester (Succinimidyl Ester) - FAQs
Alexa Fluor™ 350 NHS Ester (Succinimidyl Ester) - FAQs
View additional product information for Alexa Fluor™ 350 NHS Ester (Succinimidyl Ester) - FAQs (A10168)
3 product FAQs found
I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?
Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.
I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?
This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.
What are the signal intensity differences between Alexa Fluor 350 dye and Alexa Fluor 488 dye?
In general, blue fluorescent dyes are not as bright as other dyes further along the color spectrum. Blue dyes are structurally smaller and have lower extinction coefficients, so they are typically not as bright compared to the green, red, and far red dyes.
When using an Alexa Fluor 350 secondary antibody, we recommend that you use it for highly expressed targets and at a higher concentration than what is typically required for green or red secondary antibodies.