Sodium perborate tetrahydrate, 97%
Sodium perborate tetrahydrate, 97%
Sodium perborate tetrahydrate, 97%
Sodium perborate tetrahydrate, 97%
Thermo Scientific Chemicals

Sodium perborate tetrahydrate, 97%

CAS: 10486-00-7 | NaBO3·4H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10189.22
also known as A10189-22
Price (USD)
44.40
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
44.40
Each
Add to cart
Chemical Identifiers
CAS10486-00-7
IUPAC Namedisodium tetrahydroxy-1,2,4,5,3,6-tetraoxadiborinane-3,6-diuide hexahydrate
Molecular FormulaB2H16Na2O14
InChI KeyMCYPYXIRGFEFGL-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Na+].[Na+].O[B-]1(O)OO[B-](O)(O)OO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Redox Titration)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder or granules
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium perborate tetrahydrate is considered as a source of active hydrogen used in detergents, cleaning products, laundry detergents and bleaches. It finds application as a tooth bleaching agent utilized for tooth. It has antiseptic properties and acts as disinfectant as well as preservative in eye drops. Further, it is mixed with suitable activator, tetraacetylethylenediamine in order to release oxygen at lower temperatures.

Solubility
Soluble in water.

Notes
Moisture sensitive.Incompatible with metals, metallic salts, acids, bases and reducing agents.
RUO – Research Use Only

General References:

  1. Useful, inexpensive oxidant. Reviews: Chemtech, 566 (1991); Tetrahedron, 51, 6145 (1995); Synthesis, 1325 (1995); J. Chem. Soc., Perkin 1, 471 (2000). Examples: Oxidation of secondary alcohols to ketones, and ɑ-hydroxy acids to keto acids: Synthesis, 765 (1989). Mild epoxidation of ɑß-unsaturated ketones: Synth. Commun., 19, 3579 (1989); Tetrahedron Lett., 36, 663 (1995). Partial hydrolysis of nitriles to amides: Tetrahedron, 45, 3299 (1989); Synth. Commun., 20, 563 (1990). Oxidation (in AcOH) of sulfides to sulfoxides or sulfones, arylamines to nitro compounds, and Baeyer-Villiger oxidation of ketones to esters: Tetrahedron, 43, 1753 (1987). Oxidation (in AcOH) of aromatic aldehydes to acids: Tetrahedron, 45, 3299 (1989); or oximes to nitro compounds: Synlett, 337 (1992). Oxidative conversion of organoboranes to alcohols: Org. Synth. Coll., 9, 522 (1998).
  2. In combination with triflic acid for hydroxylation of aromatics: Synlett, 39 (1991).
  3. In combination with acetic anhydride, cyclic acetals are cleaved to their respective esters: Synlett., 329 (1995).
  4. For phase-transfer oxidation of alcohols to ketones, catalyzed by Chromium(VI) oxide, 12522, see: Synth. Commun., 21, 575 (1991).
  5. For a brief feature on uses in synthesis, see: Synlett, 2513 (2006).
  6. Qin, S.; Yin, B.; Zhang, Y.; Zhang, Y. Leaching kinetics of szaibelyite ore in NaOH solution. Hydrometallurgy 2015, 157, 333-339.
  7. Hematian, S.; Hormozi, F. Drying kinetics of coated sodium percarbonate particles in a conical fluidized bed dryer. Powder Technol. 2015, 269, 30-37.