2-Amino-4-chlorobenzoic acid, 98%
2-Amino-4-chlorobenzoic acid, 98%
2-Amino-4-chlorobenzoic acid, 98%
Thermo Scientific Chemicals

2-Amino-4-chlorobenzoic acid, 98%

CAS: 89-77-0 | C7H6ClNO2 | 171.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
100 g
250 g
Catalog number A10201.22
also known as A10201-22
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS89-77-0
IUPAC Name2-amino-4-chlorobenzoic acid
Molecular FormulaC7H6ClNO2
InChI KeyJYYLQSCZISREGY-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only