4-Bromo-m-xylene, 97%
4-Bromo-m-xylene, 97%
4-Bromo-m-xylene, 97%
Thermo Scientific Chemicals

4-Bromo-m-xylene, 97%

CAS: 583-70-0 | C8H9Br | 185.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A10212.14
also known as A10212-14
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.90
Save 7.25 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.90
Save 7.25 (15%)
Each
Chemical Identifiers
CAS583-70-0
IUPAC Name1-bromo-2,4-dimethylbenzene
Molecular FormulaC8H9Br
InChI KeyYSFLQVNTBBUKEA-UHFFFAOYSA-N
SMILESCC1=CC=C(Br)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5490-1.5540 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only