2-Acetylbenzoic acid, 98+%
2-Acetylbenzoic acid, 98+%
2-Acetylbenzoic acid, 98+%
Thermo Scientific Chemicals

2-Acetylbenzoic acid, 98+%

CAS: 577-56-0 | C9H8O3 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10216.09
also known as A10216-09
Price (USD)
53.65
Online Exclusive
59.40
Save 5.75 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.40
Save 5.75 (10%)
Each
Chemical Identifiers
CAS577-56-0
IUPAC Name2-acetylbenzoic acid
Molecular FormulaC9H8O3
InChI KeyQDAWXRKTSATEOP-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream or pale yellow to pale orange
Melting Point (clear melt)114.0-120.0?C
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.

Solubility
Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.

Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

General References:

  1. Jacob Finkelstein; Thomas Williams; Voldemar Toome; Samuel Traiman. Ring-chain tautomers of 6-substituted 2-acetylbenzoic acids. J. Org. Chem. 1967, 32 (10), 3229-3230.
  2. A. J. Dobson and R. E. Gerkin. 2-Acetylbenzoic Acid: Phthalide Form. Acta Cryst. 1996, C52 3078-3081.
  3. Exists in equilibrium with 3-hydroxy-3-methylphthalide, precursor of 3-methylenephthalidyl (MPID) derivatives of amino acids. Deprotection can be effected with hydrazine hydrate: Synthesis, 43 (1977).