Atropine sulfate monohydrate, 97+%
Atropine sulfate monohydrate, 97+%
Atropine sulfate monohydrate, 97+%
Thermo Scientific Chemicals

Atropine sulfate monohydrate, 97+%

CAS: 5908-99-6 | C17H27NO8S | 405.462 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10236.09
also known as A10236-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS5908-99-6
IUPAC Name8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate sulfuric acid hydrate
Molecular FormulaC17H27NO8S
InChI KeyPVGPXGKNDGTPTD-UHFFFAOYNA-N
SMILESO.OS(O)(=O)=O.CN1C2CCC1CC(C2)OC(=O)C(CO)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
Identification (FTIR)Conforms
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.

Solubility
Soluble in water (2500 mg/ml) at 4°C, ethanol (200 mg/ml) at 4°C, Glycerol (400 mg/ml), and methanol.

Notes
Light Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Tsung-Ming Shih; John H. McDonough JR. Organophosphorus Nerve Agents-Induced Seizures and Efficacy of Atropine Sulfate as Anticonvulsant Treatment. Pharmacology Biochemistry and Behavior. 2000, 64, (1), 147-153.
  2. C Kirchhoff; Y Bitar; S Ebel; U Holzgrabe. Analysis of atropine, its degradation products and related substances of natural origin by means of reversed-phase high-performance liquid chromatographyJ. Chromatogr. A. 2004, 1046, (1-2), 115-120.