2,2',4,4'-Tetrahydroxybenzophenone, 98+%
2,2',4,4'-Tetrahydroxybenzophenone, 98+%
2,2',4,4'-Tetrahydroxybenzophenone, 98+%
2,2',4,4'-Tetrahydroxybenzophenone, 98+%
Thermo Scientific Chemicals

2,2',4,4'-Tetrahydroxybenzophenone, 98+%

CAS: 131-55-5 | C13H10O5 | 246.218 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A10237.14
also known as A10237-14
Price (USD)
43.65
Online Exclusive
48.10
Save 4.45 (9%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.10
Save 4.45 (9%)
Each
Add to cart
Chemical Identifiers
CAS131-55-5
IUPAC Name4-(2,4-dihydroxybenzoyl)benzene-1,3-diol
Molecular FormulaC13H10O5
InChI KeyWXNRYSGJLQFHBR-UHFFFAOYSA-N
SMILESOC1=CC=C(C(=O)C2=CC=C(O)C=C2O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Identification (FTIR)Conforms
FormPowder
Assay (HPLC)≥98.0%
2,2',4,4'-Tetrahydroxybenzophenone is used as the raw material of ultraviolet absorbers in cosmetics, plastics, resins and rubber. It is used as pharmaceutical intermediates and light-sensitive materials. It is also used as pharmaceutical adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′,4,4′-Tetrahydroxybenzophenone is used as the raw material of ultraviolet absorbers in cosmetics, plastics, resins and rubber. It is used as pharmaceutical intermediates and light-sensitive materials. It is also used as pharmaceutical adjuvant.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

General References:

  1. Armin Zenker; Hansruedi Schmutz; Karl Fent. Simultaneous trace determination of nine organic UV-absorbing compounds (UV filters) in environmental samples. Journal of Chromatography A. 2008, 1202 (1), 64-74.
  2. Petra Y Kunz; Karl Fent. Estrogenic activity of UV filter mixtures. Toxicology and Applied Pharmacology. 2006, 217 (1), 86-99.