Biphenyl, 99%
Biphenyl, 99%
Biphenyl, 99%
Thermo Scientific Chemicals

Biphenyl, 99%

CAS: 92-52-4 | C12H10 | 154.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10265.0B
also known as A10265-0B
Price (USD)
38.50
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
38.50
Each
Chemical Identifiers
CAS92-52-4
IUPAC Name1,1'-biphenyl
Molecular FormulaC12H10
InChI KeyZUOUZKKEUPVFJK-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder or flakes
Melting Point (clear melt)67.0-73.0?C
Identification (FTIR)Conforms
Assay (GC)≥98.5%
Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds

Solubility
Soluble in alcohol and ether

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. keep container tightly closed.
RUO – Research Use Only

General References:

  1. J. Trotter.The crystal and molecular structure of biphenyl. Acta Cryst. 1961, 14,1135-114.
  2. Detlef E. Schulz; Gert Petrick; Jan C. Duinker.Complete characterization of polychlorinated biphenyl congeners in commercial Aroclor and Clophen mixtures by multidimensional gas chromatography-electron capture detection. Environ. Sci. Technol. 1989, 23, (7), 852-859.
  3. For use in combination with Li metal for the generation of activated Ca metal, see: Org. Synth. Coll., 9, 9 (1998).