3-Chlorobenzenesulfonyl chloride, 98%
3-Chlorobenzenesulfonyl chloride, 98%
3-Chlorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

3-Chlorobenzenesulfonyl chloride, 98%

CAS: 2888-06-4 | C6H4Cl2O2S | 211.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10283.14
also known as A10283-14
Price (USD)
288.65
Special offer
Online exclusive
Ends: 28-Dec-2026
339.00
Save 50.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
288.65
Special offer
Online exclusive
Ends: 28-Dec-2026
339.00
Save 50.35
Each
Chemical Identifiers
CAS2888-06-4
IUPAC Name3-chlorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4Cl2O2S
InChI KeyOINWZUJVEXUHCC-UHFFFAOYSA-N
SMILESClC1=CC=CC(=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormFused solid or clear liquid as melt
Appearance (Color)Colorless to pale yellow
Refractive Index1.5665-1.5705 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only