Ethyl isovalerate, 98%
Ethyl isovalerate, 98%
Ethyl isovalerate, 98%
Thermo Scientific Chemicals

Ethyl isovalerate, 98%

CAS: 108-64-5 | C7H14O2 | 130.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A10292.AE
also known as A10292-AE
Price (USD)
24.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.40
Save 3.75 (13%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
24.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.40
Save 3.75 (13%)
Each
Chemical Identifiers
CAS108-64-5
IUPAC Nameethyl 3-methylbutanoate
Molecular FormulaC7H14O2
InChI KeyPPXUHEORWJQRHJ-UHFFFAOYSA-N
SMILESCCOC(=O)CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3955-1.3995 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only