2-Chlorophenol, 99%
2-Chlorophenol, 99%
2-Chlorophenol, 99%
Thermo Scientific Chemicals

2-Chlorophenol, 99%

CAS: 95-57-8 | C6H5ClO | 128.555 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A10311.30
also known as A10311-30
Price (USD)
36.80
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
36.80
Each
Chemical Identifiers
CAS95-57-8
IUPAC Name2-chlorophenol
Molecular FormulaC6H5ClO
InChI KeyISPYQTSUDJAMAB-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (Silylated GC)≥98.5%
Refractive Index1.5580-1.5610 @ 20?C
Appearance (Color)Clear colorless to pale yellow or pink
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only