3-Aminophthalic acid hydrochloride dihydrate, 96%
Thermo Scientific Chemicals

3-Aminophthalic acid hydrochloride dihydrate, 96%

CAS: 1852533-96-0 | C8H8ClNO4 | 217.605 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10319.03
also known as A10319-03
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.70
Save 6.05
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.70
Save 6.05
Each
Chemical Identifiers
CAS1852533-96-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale grey to pale pink
FormPowder
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only