Magnesium nitrate hexahydrate, 98%
Magnesium nitrate hexahydrate, 98%
Magnesium nitrate hexahydrate, 98%
Thermo Scientific Chemicals

Magnesium nitrate hexahydrate, 98%

CAS: 13446-18-9 | H12MgN2O12 | 256.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10329.0B
also known as A10329-0B
Price (USD)
65.65
Online exclusive
73.00
Save 7.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
65.65
Online exclusive
73.00
Save 7.35
Each
Chemical Identifiers
CAS13446-18-9
IUPAC Namemagnesium(2+) hexahydrate dinitrate
Molecular FormulaH12MgN2O12
InChI KeyMFUVDXOKPBAHMC-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or compacted powder
Complexometric Titration≥97.5 to ≤102.5%
Appearance (Color)White

It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

Solubility
Very soluble in water, alcohol and liquid ammonia.

Notes
Heating of magnesium nitrate hexahydrate does not lead to dehydration but decomposition to produce magnesium oxide, nitrogen oxide and oxygen.
RUO – Research Use Only

General References:

  1. Sharma, A.; Tyagi, V. V.; Chen, C. R.; Buddhi, D. Review on thermal energy storage with phase change materials and applications. Renewable Sustainable Energy Rev. 2009, 13, 318-345.
  2. Sulaiman, M.; Rahman, A. A.; Mohamed, N. S. Structural, Thermal and Conductivity Studies of Magnesium Nitrate - Alumina Composite Solid Electrolytes Prepared via Sol-Gel Method. Int. J. Electrochem. Sci. 2013, 8, 6647 - 6655.