2-(4-Hydroxyphenylazo)benzoic acid, 97%
2-(4-Hydroxyphenylazo)benzoic acid, 97%
2-(4-Hydroxyphenylazo)benzoic acid, 97%
Thermo Scientific Chemicals

2-(4-Hydroxyphenylazo)benzoic acid, 97%

CAS: 1634-82-8 | C13H10N2O3 | 242.234 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A10333.09
also known as A10333-09
Price (USD)
58.10
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
58.10
Each
Chemical Identifiers
CAS1634-82-8
IUPAC Name2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzoic acid
Molecular FormulaC13H10N2O3
InChI KeyFBVSMDPNVYJNON-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1NN=C1C=CC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1.0%
Appearance (Color)Yellow to orange to brown
View more
Indicator used in determination of serum albumin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only