N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%
Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%

CAS: 536-46-9 | C8H14Cl2N2 | 209.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10343.22
also known as A10343-22
Price (USD)
252.65
Online Exclusive
280.00
Save 27.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
252.65
Online Exclusive
280.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS536-46-9
IUPAC NameN1,N1-dimethylbenzene-1,4-diamine dihydrochloride
Molecular FormulaC8H14Cl2N2
InChI KeyIAEDWDXMFDKWFU-UHFFFAOYSA-N
SMILESCl.Cl.CN(C)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMaterial may darken on storage
CommentMay contain a small quantity of discolored material from the manufacturing process
Appearance (Color)White to brown or dark purple to purple to pink to dark gray to gray
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
View more
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.

Solubility
Soluble in water

Notes
Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light. Store under inert gas.
RUO – Research Use Only

General References:

  1. Kazuo Hirayama and Nobuyuki Unohara. Spectrophotometric catalytic determination of an ultratrace amount of iron(III) in water based on the oxidation of N,N-dimethyl-p-phenylenediamine by hydrogen peroxide.Anal. Chem.,1988,60(23), 2573-2577.
  2. Leland L. Smith and Freddie L. Hill. Detection of sterol hydroperoxides on thin-layer chromatoplates by means of the wurster dyes.J. Chromatogr. A,1972,66(1), 101-109.