Benzyltri-n-butylammonium chloride, 98%
Benzyltri-n-butylammonium chloride, 98%
Benzyltri-n-butylammonium chloride, 98%
Thermo Scientific Chemicals

Benzyltri-n-butylammonium chloride, 98%

CAS: 23616-79-7 | C19H34ClN | 311.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A10345.0E
also known as A10345-0E
Price (USD)
362.65
Online Exclusive
403.00
Save 40.35 (10%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
362.65
Online Exclusive
403.00
Save 40.35 (10%)
Each
Chemical Identifiers
CAS23616-79-7
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only