2-Thiopheneacetic acid, 98%
2-Thiopheneacetic acid, 98%
2-Thiopheneacetic acid, 98%
Thermo Scientific Chemicals

2-Thiopheneacetic acid, 98%

CAS: 1918-77-0 | C6H6O2S | 142.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10394.14
also known as A10394-14
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.90
Save 8.25
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.90
Save 8.25
Each
Chemical Identifiers
CAS1918-77-0
IUPAC Name2-(thiophen-2-yl)acetic acid
Molecular FormulaC6H6O2S
InChI KeySMJRBWINMFUUDS-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms (non-U.S. sourced material)
CommentMaterial sourced in the U.S. and in other countries
Melting Point (clear melt)61-66°C
Appearance (Color)White to grey or brown
View more
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.

Solubility
Soluble in water, ethanol, ether and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Bae, S. E.; Kim, K. J.; Hwang, Y. K.; Huh, S. Simple preparation of Pd-NP/polythiophene nanospheres for heterogeneous catalysis. J. Colloid Interface Sci. 2015, 456, 93-99.
  2. Chen, X.; Chen, T.; Ji, F.; Zhou, Y.; Yin, S. F. Iron-catalyzed aerobic oxidative functionalization of sp3 C-H bonds: a versatile strategy for the construction of N-heterocycles. Catal. Sci. Technol. 2015, 5 (4), 2197-2202.