3-Methylthio-1-propanol, 98%
3-Methylthio-1-propanol, 98%
3-Methylthio-1-propanol, 98%
Thermo Scientific Chemicals

3-Methylthio-1-propanol, 98%

CAS: 505-10-2 | C4H10OS | 106.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10403.22
also known as A10403-22
Price (USD)
377.65
Online Exclusive
419.00
Save 41.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
377.65
Online Exclusive
419.00
Save 41.35 (10%)
Each
Chemical Identifiers
CAS505-10-2
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4895-1.4935 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only