4-Pentyn-1-ol, 95%
4-Pentyn-1-ol, 95%
4-Pentyn-1-ol, 95%
Thermo Scientific Chemicals

4-Pentyn-1-ol, 95%

CAS: 5390-04-5 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10405.06
also known as A10405-06
Price (USD)
49.65
Online Exclusive
55.50
Save 5.85 (11%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
55.50
Save 5.85 (11%)
Each
Chemical Identifiers
CAS5390-04-5
IUPAC Namepent-4-yn-1-ol
Molecular FormulaC5H8O
InChI KeyCRWVOXFUXPYTRK-UHFFFAOYSA-N
SMILESOCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥94.0%
Water Content (Karl Fischer Titration)≤1.5%
Refractive Index1.4425-1.4475 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Pentyn-1-ol is widely utilized in ring-opening polymerization reactions. It plays an important role as a starting reagent in stereo selective total synthesis of antimicrobial marine metabolites, ieodomycin A and B. It is also used in the preparation of 3-pent-4-ynyloxy phthalonitrile.

Solubility
Miscible with water.

Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Readily forms a dilithio-derivative, allowing regioselective C-alkylation in the 5-position without the need for protection of the alcohol function. Thus, alkylation with the THP ether of 5-bromopentanol gives a 10-carbon chain as part of the synthesis of a macrolide pheromone, and in other natural product syntheses: J. Org. Chem., 58, 7535 (1993), and references therein.
  2. Fjermestad, T.; Ho, J. H. H.; Macgregor, S. A.; Messerle, B. A.; Tuna, D. Computational Study of the Mechanism of Cyclic Acetal Formation via the Iridium(I)-Catalyzed Double Hydroalkoxylation of 4-Pentyn-1-ol with Methanol. Organometallics 2011, 30 (3), 618-626.
  3. Møllendal, H.; Dreizler, H.; Sutter, D. H. Structural and Conformational Properties of 4-Pentyn-1-ol As Studied by Microwave Spectroscopy and Quantum Chemical Calculations. J. Phys. Chem. A 2007, 111 (46), 11801-11808.