alpha-Methylcinnamaldehyde, predominantly (E), 97%
alpha-Methylcinnamaldehyde, predominantly (E), 97%
alpha-Methylcinnamaldehyde, predominantly (E), 97%
Thermo Scientific Chemicals

alpha-Methylcinnamaldehyde, predominantly (E), 97%

CAS: 101-39-3 | C10H10O | 146.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A10429.36
also known as A10429-36
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.90
Save 12.25 (14%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.90
Save 12.25 (14%)
Each
Chemical Identifiers
CAS101-39-3
IUPAC Name(2Z)-2-methyl-3-phenylprop-2-enal
Molecular FormulaC10H10O
InChI KeyVLUMOWNVWOXZAU-CLFYSBASSA-N
SMILESCC(C=O)=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
Free acid (titration)≤1.5%
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.6030-1.6080 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only