Cholesteryl chloride, 98%, Thermo Scientific Chemicals
Cholesteryl chloride, 98%, Thermo Scientific Chemicals
Cholesteryl chloride, 98%, Thermo Scientific Chemicals
Cholesteryl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cholesteryl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A10433.14
also known as A10433-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS910-31-6
IUPAC Name(1R,3aS,3bS,7S,9aR,9bS,11aR)-7-chloro-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene
Molecular FormulaC27H45Cl
InChI KeyOTVRYZXVVMZHHW-DPAQBDIFSA-N
SMILESCC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](Cl)CC[C@]4(C)[C@H]3CC[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-26.0 ± 1.0? (C=1 in benzene)
Appearance (Color)White
Purity (DSC)≥97.5%
Melting Point (clear melt)92.0-99.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only