2-Amino-6-methylpyridine, 99%
2-Amino-6-methylpyridine, 99%
2-Amino-6-methylpyridine, 99%
2-Amino-6-methylpyridine, 99%
Thermo Scientific Chemicals

2-Amino-6-methylpyridine, 99%

CAS: 1824-81-3 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A10444.36
also known as A10444-36
Price (USD)
140.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
140.00
Each
Add to cart
Chemical Identifiers
CAS1824-81-3
IUPAC Name6-methylpyridin-2-amine
Molecular FormulaC6H8N2
InChI KeyQUXLCYFNVNNRBE-UHFFFAOYSA-N
SMILESCC1=CC=CC(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline or fused solid
Water Content (Karl Fischer Titration)<1.0%
Assay (GC)≥98.5%
Appearance (Color)White to yellow or pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only