Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine
Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine
Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine
Thermo Scientific Chemicals

Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine

CAS: 130-95-0 | C20H24N2O2 | 324.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10459.09
also known as A10459-09
Price (USD)
71.10
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
71.10
Each
Chemical Identifiers
CAS130-95-0
IUPAC Name(R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
Molecular FormulaC20H24N2O2
InChI KeyLOUPRKONTZGTKE-ZCXAXGJJNA-N
SMILES[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMay contain up to 5% dihydroquinine (non-U.S. sourced material)
CommentSpecification differs for U.S. and non-U.S. material where indicated
FormPowder
Assay (Non-aqueous acid-base Titration)≥98.5% (non-U.S. sourced material)
Assay from Suppliers CofA≥98.5% (U.S. sourced material)
View more
Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.

Solubility
Soluble in alcohol, chloroform and diethyl ether. Slightly soluble in water and glycerol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Porta, R.; Benaglia, M.; Coccia, F.; Rossi, S.; Puglisi, A. Enantioselective Organocatalysis in Microreactors: Continuous Flow Synthesis of a (S)-Pregabalin Precursor and (S)-Warfarin. Symmetry 2015, 7 (3), 1395-1409.
  2. Porta, R.; Coccia, F.; Annunziata, R.; Puglisi, A. Comparison of Different Polymer-and Silica-Supported 9-Amino-9-deoxy-epi-quinines as Recyclable Organocatalysts. ChemCatChem 2015, 7 (9), 1490-1499.
  3. Kessler, S.; González, J.; Vlimant, M.; Glauser, G.; Guerin, P. M. Quinine and artesunate inhibit feeding in the African malaria mosquito Anopheles gambiae: the role of gustatory organs within the mouthparts. Physiol. Entomol. 2014, 39 (2), 172-182.