Glutaraldehyde, 50% aq. soln.
Glutaraldehyde, 50% aq. soln.
Glutaraldehyde, 50% aq. soln.
Thermo Scientific Chemicals

Glutaraldehyde, 50% aq. soln.

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A10500.22
also known as A10500-22
Price (USD)
33.90
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
33.90
Each
Chemical Identifiers
CAS111-30-8
IUPAC Namepentanedial
Molecular FormulaC5H8O2
InChI KeySXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILESO=CCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (Titration)≥48.0%
CommentMay contain up to 10% methanol
Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

Solubility
Miscible with water, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.

Notes
Incompatible with strong oxidisng agents, strong acids and strong bases.
RUO – Research Use Only

General References:

  1. Woods, A.; Patela, A.; Spina, D.; Riffo-Vasquez, Y.; Babin-Morgan, A.; Rosales, R. T. M.; Sunassee, K.; Clark, S.; Collins, H.; Bruce, K.; Dailey, L. A.; Forbes, B. In vivo biocompatibility, clearance, and biodistribution of albumin vehicles for pulmonary drug delivery. J. Controlled Release 2015, 210, 1-9.
  2. Lu, W.; Ma, M.; Xu, H.; Zhang, B.; Cao, X.; Guo, Y. Gelatin nanofibers prepared by spiral-electrospinning and cross-linked by vapor and liquid-phase glutaraldehyde. Mater. Lett. 2015, 140, 1-4.