Benzamide, 98+%
Benzamide, 98+%
Benzamide, 98+%
Thermo Scientific Chemicals

Benzamide, 98+%

CAS: 55-21-0 | C7H7NO | 121.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A10501.0E
also known as A10501-0E
Price (USD)
358.65
Online Exclusive
398.00
Save 39.35 (10%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
358.65
Online Exclusive
398.00
Save 39.35 (10%)
Each
Chemical Identifiers
CAS55-21-0
IUPAC Namebenzamide
Molecular FormulaC7H7NO
InChI KeyKXDAEFPNCMNJSK-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting Point (clear melt)125-129°C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Benzamide is utilized to study the mechanism of photocatalytic decomposition of aqueous solution of acetic acid, acetamide and acetonitrile in the presence of semiconductors. It is used as a nictoinamide-mimic PARP inhibitor and neuroprotectant. Further, it is used to develop a robust screening method to study biotransformations using (+)-gamma-lactamase enzyme. It is also employed in the determination of glycine. In addition to this, it is used as an intermediate in organic synthesis as well as in the production of pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzamide is utilized to study the mechanism of photocatalytic decomposition of aqueous solution of acetic acid, acetamide and acetonitrile in the presence of semiconductors. It is used as a nictoinamide-mimic PARP inhibitor and neuroprotectant. Further, it is used to develop a robust screening method to study biotransformations using (+)-gamma-lactamase enzyme. It is also employed in the determination of glycine. In addition to this, it is used as an intermediate in organic synthesis as well as in the production of pharmaceuticals and dyes.

Solubility
Soluble in water, methanol, ammonia, pyridine, carbon tetrachloride, carbon disulfide and ethanol. Slightly soluble in benzene and ethyl ether.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Reduction with lithium aluminum hydride gives benzylamine. By using lithium tris(diethylamino)aluminum hydride (see Diethyl amine, A11716), selective reduction to benzaldehyde is possible: Tetrahedron Lett., 32, 6903 (1991).
  2. An exceptionally mild method for dehydration of amides to nitriles by transamidation with acetonitrile in the presence of an aldehyde and formic acid is exemplified by the high yield conversion of benzamide to benzonitrile: J. Org. Chem., 61, 6486 (1996).
  3. Barrett, K. T.; Miller, S. J. Regioselective Derivatizations of a Tribrominated Atropisomeric Benzamide Scaffold. Org. Lett. 2015, 17 (3), 580-583.
  4. Manna, S.; Antonchick, A. P. Organocatalytic Oxidative Annulation of Benzamide Derivatives with Alkynes. Angew. Chem. Int. Ed. 2014, 53 (28), 7324-7327.