Product Image
Applied Biosystems™

SequalPrep™ Wash Buffer

SequalPrep™ Wash Buffer
Have Questions?
Catalog NumberQuantity
A1051003
also known as A10510-03
Catalog number A1051003
also known as A10510-03
Price (USD)
1,114.00
Each
Price (USD)
1,114.00
Each

For use with SequalPrep™ Normalization Plates, the wash buffer may be purchased separately.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Product LineSequalPrep
Product TypeWash Buffer
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.