Can I use the GFP BrightComp eBeads to compensate for FITC or Alexa Fluor 488?
No. You must always compensate with the same fluorophore present on the samples. The GFP BrightComp eBeads Compensation Bead Kit spectrally emulates GFP and must only be used on GFP-expressing samples.