Pentaerythritol, 98+%
Pentaerythritol, 98+%
Pentaerythritol, 98+%
Pentaerythritol, 98+%
Thermo Scientific Chemicals

Pentaerythritol, 98+%

CAS: 115-77-5 | C5H12O4 | 136.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10520.0B
also known as A10520-0B
Price (USD)
41.65
Online Exclusive
45.70
Save 4.05 (9%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
45.70
Save 4.05 (9%)
Each
Add to cart
Chemical Identifiers
CAS115-77-5
IUPAC Name2,2-bis(hydroxymethyl)propane-1,3-diol
Molecular FormulaC5H12O4
InChI KeyWXZMFSXDPGVJKK-UHFFFAOYSA-N
SMILESOCC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (Silylated GC)≥98.0%
Water Content (Karl Fischer Titration)≤1.0%
Melting Point257-265?C
View more

Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

Solubility
Soluble in Water: 0.1 g/mL

Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents, acid chlorides, acid anhydrides.
RUO – Research Use Only
James J. Savage; Robert H. Wood. Enthalpy of dilution of aqueous mixtures of amides, sugars, urea, ethylene glycol, and pentaerythritol at 25°C: Enthalpy of interaction of the hydrocarbon, amide, and hydroxyl functional groups in dilute aqueous solutions. Journal of Solution Chemistry..1976, 5, 733-750.