What storage conditions should I use for Lithium chloride?
Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.
Search
View additional product information for Tetramethylrhodamine Goat Anti-Rat IgG (H+L) - FAQs (A10531)
1 product FAQs found