4,6-Dihydroxy-2-methylpyrimidine, 99%
4,6-Dihydroxy-2-methylpyrimidine, 99%
4,6-Dihydroxy-2-methylpyrimidine, 99%
Thermo Scientific Chemicals

4,6-Dihydroxy-2-methylpyrimidine, 99%

CAS: 40497-30-1 | C5H6N2O2 | 126.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A10535.18
also known as A10535-18
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS40497-30-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale pink
Identification (FTIR)Conforms
Assay (Silylated GC)≥98.5%
FormPowder
4,6-Dihydroxy-2-methylpyrimidine is used in the preparation of 4,6-dichloro-2-methyl-pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dihydroxy-2-methylpyrimidine is used in the preparation of 4,6-dichloro-2-methyl-pyrimidine.

Solubility
Soluble in sodium hydroxide.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhang, J.; Zhang, Q.; Vo, T. T.; Parrish, D. A.; Shreeve, J. N. M. Energetic Salts with pi-Stacking and Hydrogen-Bonding Interactions Lead the Way to Future Energetic Materials. J. Am. Chem. Soc. 2015, 137 (4), 1697-1704.
  2. Saikia, A.; Sivabalan, R.; Gore, G. M.; Sikder, A. K. Synthesis of some Potential High Energy Materials using Metal Nitrates; An approach towards Environmental Benign Process. J. Sci. Ind. Res. 2014, 73 (7), 485-488.