Sodium bromide, 99+% (dry wt.), water <1.0%
Sodium bromide, 99+% (dry wt.), water <1.0%
Sodium bromide, 99+% (dry wt.), water <1.0%
Sodium bromide, 99+% (dry wt.), water <1.0%
Thermo Scientific Chemicals

Sodium bromide, 99+% (dry wt.), water <1.0%

CAS: 7647-15-6 | BrNa | 102.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A10552.0I
also known as A10552-0I
Price (USD)
338.65
Online Exclusive
376.00
Save 37.35 (10%)
Each
Quantity:
5000 g
Request bulk or custom format
Price (USD)
338.65
Online Exclusive
376.00
Save 37.35 (10%)
Each
Chemical Identifiers
CAS7647-15-6
IUPAC Namesodium bromide
Molecular FormulaBrNa
InChI KeyJHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES[Na+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Titration ex Bromide)≥99.0 to ≤101.0% (dry wt. basis)
Water Content (Karl Fischer Titration)≤1%
Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

Solubility
Soluble in water, soluble in alcohol.

Notes
It is thermally and chemically stable. Hygroscopic. Sodium bromide is non-flammable and when strongly heated it may emit bromine fumes.
RUO – Research Use Only

General References:

  1. For use in a facile method for conversion of primary alkyl chlorides to bromides, see: Synth. Commun., 14, 1313 (1984). In combination with TMS chloride and bromine in 2:1 methanol-acetonitrile, provides a method for the ɑ -bromination of dimethyl acetals: Gazz. Chim. Ital., 123, 629 (1993).
  2. Srinivasa Rao, K.; Gehlot, P. S.; Gupta, H.; Drechsler, M.; Kumar, A. Sodium Bromide Induced Micelle to Vesicle Transitions of Newly Synthesized Anionic Surface Active Ionic Liquids Based on Dodecylbenzenesulfonate. J. Phys. Chem. B 2015, 119, (11), 4263-4274.
  3. Wang, G. W.; Gao, J. Solvent-free bromination reactions with sodium bromide and oxone promoted by mechanical milling. Green Chem. 2012, 14, (4), 1125-1131.