4-Bromothiophenol, 98%
4-Bromothiophenol, 98%
4-Bromothiophenol, 98%
Thermo Scientific Chemicals

4-Bromothiophenol, 98%

CAS: 106-53-6 | C6H5BrS | 189.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10555.30
also known as A10555-30
Price (USD)
734.65
Online Exclusive
816.00
Save 81.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
734.65
Online Exclusive
816.00
Save 81.35 (10%)
Each
Chemical Identifiers
CAS106-53-6
IUPAC Name4-bromobenzene-1-thiol
Molecular FormulaC6H5BrS
InChI KeyFTBCOQFMQSTCQQ-UHFFFAOYSA-N
SMILESSC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)71.5-77.5?C
Appearance (Color)White to pale yellow or pale brown
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused/lumpy solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only