Methyl 4-(bromomethyl)benzoate, 98%
Methyl 4-(bromomethyl)benzoate, 98%
Methyl 4-(bromomethyl)benzoate, 98%
Methyl 4-(bromomethyl)benzoate, 98%
Thermo Scientific Chemicals

Methyl 4-(bromomethyl)benzoate, 98%

CAS: 2417-72-3 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10557.06
also known as A10557-06
Price (USD)
50.65
Online Exclusive
55.80
Save 5.15 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
55.80
Save 5.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS2417-72-3
IUPAC Namemethyl 4-(bromomethyl)benzoate
Molecular FormulaC9H9BrO2
InChI KeyNLWBJPPMPLPZIE-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)52.0-59.0°C
Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

Solubility
Lowly soluble in water, highly soluble in ethanol and ether, not soluble in halogenated organic solvents

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. HS Yathirajan, S Bindya, BK Sarojini. Methyl 4-(bromomethyl) benzoate. Acta Cryst. 1969 E63, (1) , 1334-1335.
  2. WT Sing.; CL Lee.; SL Yeo.; SP Lim.; MM Sim. Arylalkylidene rhodanine with bulky and hydrophobic functional group as selective HCV NS3 protease inhibitor. Bioorganic & Medicinal Chemistry Letters.. 2001 11, (2) , 91-94.