N,N-Diethylaniline, 99%
N,N-Diethylaniline, 99%
N,N-Diethylaniline, 99%
Thermo Scientific Chemicals

N,N-Diethylaniline, 99%

CAS: 91-66-7 | C10H15N | 149.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
Catalog number A10560.0F
also known as A10560-0F
Price (USD)
110.65
Online Exclusive
123.00
Save 12.35 (10%)
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
110.65
Online Exclusive
123.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS91-66-7
IUPAC NameN,N-diethylaniline
Molecular FormulaC10H15N
InChI KeyGGSUCNLOZRCGPQ-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
Assay (GC)≥98.5%
Refractive Index1.5405-1.5435 @ 20?C
Identification (FTIR)Conforms
FormLiquid
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.

Solubility
Miscible with water and acetone. Slightly miscible with chloroform, alcohol and ether.      

Notes
Incompatible with strong oxidizing agents and strong acids.                  .
RUO – Research Use Only

General References:

  1. Jiang, L.; Liu, W.; Song, Y.; He, X.; Wang, Y.; Yang, Y. Photoinduced electron transfer of Rhodamine 6G, N-diethylaniline revealed by multiplex transient grating and transient absorption spectroscopies. Applied Physics B 2014, 116 (2), 271-277.
  2. Jiang, L.; Liu, W.; Song, Y.; He, X.; Wang, Y.; Wang, C.; Wu, H.; Yang, F.; Yang, Y. Photoinduced intermolecular electron transfer and off-resonance Raman characteristics of Rhodamine 101, N-diethylaniline. Chem. Phys. 2014, 429, 12-19.