1-Bromononane, 99%
1-Bromononane, 99%
1-Bromononane, 99%
Thermo Scientific Chemicals

1-Bromononane, 99%

CAS: 693-58-3 | C9H19Br | 207.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10577.22
also known as A10577-22
Price (USD)
109.65
Special offer
Online exclusive
Ends: 30-Jun-2026
129.00
Save 19.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
109.65
Special offer
Online exclusive
Ends: 30-Jun-2026
129.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS693-58-3
IUPAC Name1-bromononane
Molecular FormulaC9H19Br
InChI KeyAYMUQTNXKPEMLM-UHFFFAOYSA-N
SMILESCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4530-1.4560 @ 20?C
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

Solubility
Insoluble in water. Soluble in ethyl ether and benzene.

Notes
Stable under normal conditions. Incompatible with oxidizing agents.
RUO – Research Use Only