Isonicotinic acid hydrazide, 98+%
Isonicotinic acid hydrazide, 98+%
Isonicotinic acid hydrazide, 98+%
Thermo Scientific Chemicals

Isonicotinic acid hydrazide, 98+%

CAS: 54-85-3 | C6H7N3O | 137.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10583.0B
also known as A10583-0B
Price (USD)
191.65
Online exclusive
213.00
Save 21.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
191.65
Online exclusive
213.00
Save 21.35
Each
Chemical Identifiers
CAS54-85-3
IUPAC Namepyridine-4-carbohydrazide
Molecular FormulaC6H7N3O
InChI KeyQRXWMOHMRWLFEY-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystals
Assay from Suppliers CofA≥98.0% (GC, U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Elemental AnalysisC: 52.55 ± 0.6% (non-U.S. specification)
View more
Used for fluorescent HPLC detection of delta 4-3-ketosteroids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only