1-Bromododecane, 98%
1-Bromododecane, 98%
1-Bromododecane, 98%
Thermo Scientific Chemicals

1-Bromododecane, 98%

CAS: 143-15-7 | C12H25Br | 249.236 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A10600.0B
also known as A10600-0B
Price (USD)
108.65
Online Exclusive
120.00
Save 11.35 (9%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
120.00
Save 11.35 (9%)
Each
Chemical Identifiers
CAS143-15-7
IUPAC Name1-bromododecane
Molecular FormulaC12H25Br
InChI KeyPBLNBZIONSLZBU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4560-1.4600 @ 20?C
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Giles R. Cokelet; Frederick J. Hollander; Joseph H. Smith. Density and viscosity of mixtures of 1,1,2,2-tetrabromoethane and 1-bromododecane. J. Chem. Eng. Data. 1969, 14, (4) 470-473.