Dimethyl 5-nitroisophthalate, 98+%
Dimethyl 5-nitroisophthalate, 98+%
Dimethyl 5-nitroisophthalate, 98+%
Dimethyl 5-nitroisophthalate, 98+%
Thermo Scientific Chemicals

Dimethyl 5-nitroisophthalate, 98+%

CAS: 13290-96-5 | C10H9NO6 | 239.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A10610.0B
also known as A10610-0B
Price (USD)
205.65
Special offer
Online exclusive
Ends: 30-Jun-2026
242.00
Save 36.35 (15%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
205.65
Special offer
Online exclusive
Ends: 30-Jun-2026
242.00
Save 36.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS13290-96-5
IUPAC Name1,3-dimethyl 5-nitrobenzene-1,3-dicarboxylate
Molecular FormulaC10H9NO6
InChI KeyGGTSJKFPGKFLCZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=CC(=C1)C(=O)OC)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)119.0-126.0?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only