3-Nitroaniline, 98%
3-Nitroaniline, 98%
3-Nitroaniline, 98%
Thermo Scientific Chemicals

3-Nitroaniline, 98%

CAS: 99-09-2 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A10629.36
also known as A10629-36
Price (USD)
110.65
Online Exclusive
123.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
110.65
Online Exclusive
123.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS99-09-2
IUPAC Name3-nitroaniline
Molecular FormulaC6H6N2O2
InChI KeyXJCVRTZCHMZPBD-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)110.0-116.0?C
Appearance (Color)Yellow to orange
Assay (GC)≥97.5%
3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.

Solubility
It is soluble in water 0.1 g/100 ml (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, chloroformates, strong oxidizing agents.
RUO – Research Use Only

General References:

  1. HPS Rathore.; SK Tiwari. Spectrophotometric determination of nitrite in polluted waters using 3-nitroaniline. Analytica chimica acta. 1991 242, (1) , 225-228.
  2. AS Yazdi, F Mofazzeli, Z Es'haghi. Determination of 3-nitroaniline in water samples by directly suspended droplet three-phase liquid-phase microextraction using 18-crown-6 ether and high-performance liquid chromatography. Journal of Chromatography A. 2009 1216, (26) , 5086-5091.