2-Bromo-1,3-difluorobenzene, 98%
2-Bromo-1,3-difluorobenzene, 98%
2-Bromo-1,3-difluorobenzene, 98%
Thermo Scientific Chemicals

2-Bromo-1,3-difluorobenzene, 98%

CAS: 64248-56-2 | C6H3BrF2 | 192.991 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A10633.06
also known as A10633-06
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.80
Save 7.15 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.80
Save 7.15 (14%)
Each
Chemical Identifiers
CAS64248-56-2
IUPAC Name2-bromo-1,3-difluorobenzene
Molecular FormulaC6H3BrF2
InChI KeyHRZTZLCMURHWFY-UHFFFAOYSA-N
SMILESFC1=CC=CC(F)=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow to pale brown
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5090-1.5130 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only