1-Bromo-2-fluorobenzene, 99%
1-Bromo-2-fluorobenzene, 99%
1-Bromo-2-fluorobenzene, 99%
Thermo Scientific Chemicals

1-Bromo-2-fluorobenzene, 99%

CAS: 1072-85-1 | C6H4BrF | 175 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10635.22
also known as A10635-22
Price (USD)
131.65
Special offer
Online exclusive
Ends: 30-Jun-2026
155.00
Save 23.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
131.65
Special offer
Online exclusive
Ends: 30-Jun-2026
155.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS1072-85-1
IUPAC Name1-bromo-2-fluorobenzene
Molecular FormulaC6H4BrF
InChI KeyIPWBFGUBXWMIPR-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.5325-1.5355 @ 20?C
1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents .
RUO – Research Use Only

General References:

  1. KG Rutherford.; W Redmond. 1-Bromo-2-Fluorobenzene. Organic Syntheses. 1963, 100, (1), 54-62.
  2. M Feuerstein.; H Doucet.; M Santelli. Tetraphosphine/palladium-catalysed Suzuki cross-coupling with sterically hindered aryl bromides and arylboronic acids. Tetrahedron Letters. 2001, 42, (38), 6667-6670.
  3. On treatment with Mg: J. Org. Chem., 49, 4518 (1984), or n-BuLi: Tetrahedron, 48, 4379 (1992), benzyne is generated, which undergoes [4+2] cycloadditions with furans or pyrroles.
  4. Whereas n-BuLi effects bromine-metal exchange, lithiation with LDA at -75° results in deprotonation ortho to F; subsequent reaction with electrophiles provides access to mixed dihalo derivatives such as 3-bromo-2-fluorobenzoic acid: Tetrahedron Lett., 36, 881 (1995). Improved yields are obtained by the use of lithium 2,2,6,6-tetramethylpiperidide (LTMP) as base: Tetrahedron Lett., 37, 6551 (1996).