BaculoDirect™ GST Gateway® Transfection Kit - FAQs
BaculoDirect™ GST Gateway® Transfection Kit - FAQs
View additional product information for BaculoDirect™ GST Gateway® Transfection Kit - FAQs (A10640)
1 product FAQs found
Is it normal to observe a precipitate in the ganciclovir solution after thawing?
A precipitate may sometimes appear after thawing the ganciclovir solution. If this occurs, heat the solution to 37C in a water bath for 5-10 minutes and vortex a few times. The precipitate should go back into solution. For future use, it is best to aliquot the remainder and store at -20C.